ENA413164827 Enamine

2-amino-5-chloropyridin-3-ol

  • MDL number MFCD14706179
  • Cas Number : 40966-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199-202
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113168
Availability details for
Quantity
  • Availability
  • Add to Cart