ENA413164838 Enamine

methyl 2-(hydroxymethyl)pyridine-4-carboxylate

  • MDL number MFCD11848098
  • Cas Number : 58481-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113678

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-113678
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart