ENA413164840 Enamine

methyl 4-ethyl-2-hydroxybenzoate

  • MDL number MFCD20441268
  • Cas Number : 28844-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113752

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113752
Availability details for
Quantity
  • Availability
  • Add to Cart