ENA413164849 Enamine

5,8-dimethyl-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD00060834
  • Cas Number : 5037-63-8
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114238
Availability details for
Quantity
  • Availability
  • Add to Cart