ENA413164868 Enamine

2-chloro-3-methanesulfonylpyridine

  • MDL number MFCD17016076
  • Cas Number : 70682-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-115387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115387
Availability details for
Quantity
  • Availability
  • Add to Cart