ENA413164878 Enamine

1-methyl-4-nitro-1H-imidazole-5-carbonitrile

  • MDL number MFCD00707882
  • Cas Number : 40648-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-142
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-115984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-115984
Availability details for
Quantity
  • Availability
  • Add to Cart