ENA413164880 Enamine

ethyl 5-sulfanyl-1,3,4-oxadiazole-2-carboxylate

  • MDL number MFCD26131753
  • Cas Number : 61320-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-116044

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P363,P501
hcodes Xn,Xi
MSDS https://www.enaminestore.com/catalog/EN300-116044
pictograms GHS05,GHS07
hstatements H302,H312,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart