ENA413164884 Enamine

6-amino-1H-indole-3-carboxamide

  • MDL number MFCD12790912
  • Cas Number : 1249460-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-116308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-116308
Availability details for
Quantity
  • Availability
  • Add to Cart