ENA413164929 Enamine

3,4-dihydro-2H-1,4-benzoxazine-8-carboxylic acid

  • MDL number MFCD11848184
  • Cas Number : 1007875-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-118805
Availability details for
Quantity
  • Availability
  • Add to Cart