ENA413164942 Enamine

1-(4-bromo-2-chlorophenyl)ethan-1-one

  • MDL number MFCD07781036
  • Cas Number : 252561-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-119469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-119469
Availability details for
Quantity
  • Availability
  • Add to Cart