ENA413164980 Enamine

2-[1-({[(benzyloxy)carbonyl]amino}methyl)cyclohexyl]acetic acid

  • MDL number MFCD18264502
  • Cas Number : 1083246-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121226

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121226
Availability details for
Quantity
  • Availability
  • Add to Cart