ENA413164991 Enamine

6-iodo-1,3-benzothiazol-2-amine

  • MDL number MFCD00996639
  • Cas Number : 16582-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121707

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121707
Availability details for
Quantity
  • Availability
  • Add to Cart