ENA413165037 Enamine

4,4-difluoro-1-methylcyclohexan-1-amine

  • MDL number MFCD22548294
  • Cas Number : 1389442-00-5
  • Molecular Weight : 149.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-difluoro-1-methylcyclohexan-1-amine
Inchi Code 1S/C7H13F2N/c1-6(10)2-4-7(8,9)5-3-6/h2-5,10H2,1H3
Inchi Key HMMITIYMFHIYBL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124496

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P271,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-124496
pictograms GHS02,GHS05,GHS07
hstatements H226,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart