ENA413165038 Enamine

3-methyl-1-phenylpiperazine

  • MDL number MFCD06804051
  • Cas Number : 72615-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124592
Availability details for
Quantity
  • Availability
  • Add to Cart