ENA413165063 Enamine

1-phenyl-1H-1,2,3-triazole

  • MDL number MFCD08695353
  • Cas Number : 1453-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125554

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125554
Availability details for
Quantity
  • Availability
  • Add to Cart