ENA413165070 Enamine

tert-butyl N-(2,3-dihydro-1H-indol-7-yl)carbamate

  • MDL number MFCD08234596
  • Cas Number : 885270-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125881
Availability details for
Quantity
  • Availability
  • Add to Cart