ENA413165071 Enamine

4-phenyloxolan-2-one

  • MDL number MFCD22495398
  • Cas Number : 1008-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125895

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-125895
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart