ENA413165073 Enamine

2-chloro-4-methylthiophene-3-carboxylic acid

  • MDL number MFCD20658128
  • Cas Number : 85345-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125969
Availability details for
Quantity
  • Availability
  • Add to Cart