ENA413165076 Enamine

(1S,2S)-2-methylcyclopropane-1-carboxylic acid

  • MDL number MFCD19228287
  • Cas Number : 14590-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-126145
Availability details for
Quantity
  • Availability
  • Add to Cart