ENA413165120 Enamine

methyl 3-oxo-2,3-dihydro-1H-indene-1-carboxylate

  • MDL number MFCD01846161
  • Cas Number : 29427-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-47
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129392
Availability details for
Quantity
  • Availability
  • Add to Cart