ENA413165124 Enamine

(3-methylpyridin-2-yl)methanol

  • MDL number MFCD02684206
  • Cas Number : 63071-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129623
Availability details for
Quantity
  • Availability
  • Add to Cart