ENA413165137 Enamine

methyl 2,2,3,3-tetramethylcyclopropane-1-carboxylate

  • MDL number MFCD18262384
  • Cas Number : 2415-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-103955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-103955
Availability details for
Quantity
  • Availability
  • Add to Cart