ENA413165164 Enamine

tert-butyl N-[4-(methanesulfonyloxy)cyclohexyl]carbamate

  • MDL number MFCD08272981
  • Cas Number : 692782-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130236
Availability details for
Quantity
  • Availability
  • Add to Cart