ENA413165167 Enamine

3-(4-fluorophenyl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-6-amine

  • MDL number MFCD26732412
  • Cas Number : 1551814-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130402
Availability details for
Quantity
  • Availability
  • Add to Cart