ENA413165182 Enamine

3-phenylpyrrolidine-2-carboxylic acid

  • MDL number MFCD01191420
  • Cas Number : 3005-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 275-277
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131172
Availability details for
Quantity
  • Availability
  • Add to Cart