ENA413165183 Enamine

1-(4-methoxy-2,5-dimethylphenyl)ethan-1-ol

  • MDL number MFCD14529675
  • Cas Number : 54119-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131185
Availability details for
Quantity
  • Availability
  • Add to Cart