ENA413165186 Enamine

ethyl 3-formylcyclohexane-1-carboxylate

  • MDL number MFCD00019348
  • Cas Number : 40582-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131369
Availability details for
Quantity
  • Availability
  • Add to Cart