ENA413165190 Enamine

3-{bicyclo[2.2.1]heptan-2-yl}propan-1-ol

  • MDL number MFCD21951780
  • Cas Number : 1559615-90-5
  • Molecular Weight : 154.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bicyclo[2.2.1]hept-2-yl-1-propanol
Inchi Code 1S/C10H18O/c11-5-1-2-9-6-8-3-4-10(9)7-8/h8-11H,1-7H2
Inchi Key DCYDDAKRCSSKAN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-131519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131519
Availability details for
Quantity
  • Availability
  • Add to Cart