ENA413165191 Enamine

N-cyclopropyl-4-nitroaniline

  • MDL number MFCD11122421
  • Cas Number : 112033-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131614
Availability details for
Quantity
  • Availability
  • Add to Cart