ENA413165217 Enamine

5-bromo-2-methyl-7-nitro-1,3-benzoxazole

  • MDL number MFCD16468683
  • Cas Number : 1226072-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132731

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132731
Availability details for
Quantity
  • Availability
  • Add to Cart