ENA413165222 Enamine

5,6-dimethylpyrazin-2-ol

  • MDL number MFCD13705420
  • Cas Number : 57229-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132921

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132921
Availability details for
Quantity
  • Availability
  • Add to Cart