ENA413165228 Enamine

(2-bromocyclopropyl)benzene

  • MDL number MFCD00760308
  • Cas Number : 36617-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133174
Availability details for
Quantity
  • Availability
  • Add to Cart