ENA413165238 Enamine

N-(azetidin-3-yl)-2,2,2-trifluoroacetamide hydrochloride

  • MDL number MFCD13185978
  • Cas Number : 124668-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133622
Availability details for
Quantity
  • Availability
  • Add to Cart