ENA413165243 Enamine

3-methoxypyridine-4-carbonitrile

  • MDL number MFCD13175841
  • Cas Number : 26414-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106161

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106161
Availability details for
Quantity
  • Availability
  • Add to Cart