ENA413165245 Enamine

5-phenyloxolan-3-ol

  • MDL number MFCD23889375
  • Cas Number : 197781-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106197
Availability details for
Quantity
  • Availability
  • Add to Cart