ENA413165246 Enamine

1-[2-amino-3-(4-methylbenzoyl)-4H,5H,6H-thieno[2,3-c]pyrrol-5-yl]ethan-1-one

  • MDL number MFCD26935965
  • Cas Number : 1797345-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-133877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133877
Availability details for
Quantity
  • Availability
  • Add to Cart