ENA413165254 Enamine

2-(4-bromophenyl)-1-(4-chlorophenyl)ethan-1-ol

  • MDL number MFCD12723644
  • Cas Number : 1183175-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134051
Availability details for
Quantity
  • Availability
  • Add to Cart