ENA413165261 Enamine

3-(pyridine-3-carbonyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD26935970
  • Cas Number : 1803582-68-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134229

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134229
Availability details for
Quantity
  • Availability
  • Add to Cart