ENA413165265 Enamine

ethyl 3-methyl-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD27500668
  • Cas Number : 857424-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134273

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134273
Availability details for
Quantity
  • Availability
  • Add to Cart