ENA413165266 Enamine

7-(pyrrolidin-1-yl)-1,4-dioxaspiro[4.5]decan-8-ol

  • MDL number MFCD16100714
  • Cas Number : 1248660-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134305
Availability details for
Quantity
  • Availability
  • Add to Cart