ENA413165268 Enamine

4,4,4-trifluoro-2-methyl-3-oxobutanenitrile

  • MDL number MFCD11208741
  • Cas Number : 110234-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134451
Availability details for
Quantity
  • Availability
  • Add to Cart