ENA413165270 Enamine

1-(3-amino-5-bromo-2-hydroxyphenyl)ethan-1-one

  • MDL number MFCD12196330
  • Cas Number : 70977-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101...103
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134576

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-134576
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart