ENA413165293 Enamine

3-difluoromethanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD17270946
  • Cas Number : 1247631-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106518
Availability details for
Quantity
  • Availability
  • Add to Cart