ENA413165297 Enamine

2-(cyclopropylmethyl)propanedioic acid

  • MDL number MFCD03003347
  • Cas Number : 165319-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135158

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135158
Availability details for
Quantity
  • Availability
  • Add to Cart