ENA413165307 Enamine

(1R)-1-(4-bromophenyl)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD18235544
  • Cas Number : 1259904-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135293
Availability details for
Quantity
  • Availability
  • Add to Cart