ENA413165322 Enamine

methyl 4-(1-{[(tert-butoxy)carbonyl]amino}cyclopropyl)benzoate

  • MDL number MFCD24467647
  • Cas Number : 1338243-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135752

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135752
Availability details for
Quantity
  • Availability
  • Add to Cart