ENA413165328 Enamine

N'-amino-3-methylpyridine-2-carboximidamide

  • MDL number MFCD16087884
  • Cas Number : 411210-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135859
Availability details for
Quantity
  • Availability
  • Add to Cart