ENA413165329 Enamine

2-{[(2-hydroxy-5-methylphenyl)methyl]amino}benzoic acid

  • MDL number MFCD19384925
  • Cas Number : 92841-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135867
Availability details for
Quantity
  • Availability
  • Add to Cart