ENA413165338 Enamine

N-cyclopropyl-2-methyl-6-nitroaniline

  • MDL number MFCD21098403
  • Cas Number : 1353977-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136158
Availability details for
Quantity
  • Availability
  • Add to Cart