ENA413165359 Enamine

[4-(1,1,2,2,2-pentafluoroethyl)phenyl]methanol

  • MDL number MFCD19439992
  • Cas Number : 857297-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136728
Availability details for
Quantity
  • Availability
  • Add to Cart